Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Soda lime, ACS

Catalog No. aa3659636
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3659636 500 g
AA36596A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3659636 Supplier Thermo Scientific Chemicals Supplier No. 03659636
Only null left
Add to Cart

CAS: 8006-28-8 | CaHNaO2 | 96.074 g/mol

Soda lime's ability to absorb carbon dioxide makes it valuable in medical and surgical professions. It is also used as powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil CO2 efflux upon absorption. It is also useful for direct measurement of dissolved CO2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8006-28-8
Molecular Formula CaHNaO2
Molecular Weight (g/mol) 96.074
MDL Number MFCD00134094
InChI Key HUAUNKAZQWMVFY-UHFFFAOYSA-M
PubChem CID 66545795
IUPAC Name sodium;oxocalcium;hydroxide
SMILES [OH-].O=[Ca].[Na+]

Specifications

Physical Form 4-8 Mesh
Material Soda Lime
Quantity 500 g
Mesh Size 4 to 8
Solubility Information Soluble in water.
Sensitivity Air sensitive; Hygroscopic
Grade ACS Reagent
Chemical Name or Material Soda Lime
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.