Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Soda Lime, Indicating, 4-8 Mesh, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. 18606251 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
15 kg
300 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-251 500 g Amber Glass Bottle
18-606-254 300 lb. Fiber Drum
18-606-252 2.5 kg Poly Pail
18-606-253 15 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-251 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO102500GM
Add to Cart
Add to Cart

SO102, 8006-28-8

Spectrum™ Chemical Soda Lime, Indicating, 4-8 Mesh, Reagent, ACS

Chemical Identifiers

CAS 8006-28-8
Molecular Formula CaHNaO2
Molecular Weight (g/mol) 96.07
InChI Key HUAUNKAZQWMVFY-UHFFFAOYSA-M
IUPAC Name sodium oxocalcium hydroxide
SMILES [OH-].[Na+].O=[Ca]

Specifications

CAS Max % 1
Quantity 500 g
Grade ACS
Loss on Drying 0.07
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.