Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Soda Pop Science Lab
SCI_ED

Catalog No. S67412
Change view
Click to view available options
Soda Pop Science Lab
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S67412 Soda Pop Science Lab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S67412 Supplier Neo/Sci Corporation Supplier No. 203053
This item is not returnable. View return policy

In this investigation, students dive into the study of this popular beverage.

Designed by teachers to meet key National Science Standards, including “Abilities Necessary to Do Scientific Inquiry” and “Understanding Scientific Inquiry.”

  • Gain a basic understanding of the scientific method, experimental design and basic problem-solving skills
  • Test the chemical and physical characteristics of soda
  • Discover the composition and nutritional value of various sodas

Order Info

Class size: 40 students.

Specifications

Certifications/Compliance National science standards
For Use With (Application) Students dive into the study of this popular beverage
Class Size 40 Students
Product Type Soda Pop Science Lab Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.