Learn More
Cayman Chemical Sodium Lauryl Sulfoacetate, Purity: ≥95%, CAS: 1847-58-1, 250 mg
SDP
Catalog No. 502989191
Encompass_Preferred
An anionic surfactant; increases proliferation of isolated chicken PBMCs at 62.5-500 µg/ml; increases the blood CD4+ to CD8+ T cell ratio induced by a Newcastle disease virus vaccine in chickens at 1,2, and 4 mg/kg
Catalog No. 50-298-9191 Supplier Cayman Chemical Supplier No. 29906250 MG
An anionic surfactant; increases proliferation of isolated chicken PBMCs at 62.5-500 µg/ml; increases the blood CD4+ to CD8+ T cell ratio induced by a Newcastle disease virus vaccine in chickens at 1,2, and 4 mg/kg