Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodiu mL-Tartrate Dibasic Dihydrate

Catalog No. 228729100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-872-9100G 100 g
22-872-91KG 1 kg
22-872-9500G 500 g
22-872-95G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-872-9100G Supplier Sigma Aldrich Supplier No. 228729100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodiu mL-tartrate is an organic acid mainly found in grapes.Sodiu mL-tartrate dibasic dihydrate may be used in the preparation of solid-state M-L compounds (M = bivalent metal ions ie., Mn, Fe, Co, Ni, Cu, Zn; L = tartrate). It may also be used in the preparation of copper (II) and cobalt (II) tartrate complexes by reacting with the corresponding metal salts. These complexes can undergo thermal decomposition to form Cu and Co metal oxide nanoparticles.

Specifications

CAS 6106-24-7
Molecular Formula C4H4Na2O6 -+ 2H2O
Linear Formula C4H4Na2O6 -+ 2H2O
Quantity 100 g
MDL Number MFCD00150035
Synonym Sodium tartrate dibasic dihydrate; L-(+)-Tartaric acid disodium salt; Disodium tartrate dihydrate; Sodium tartrate dihydrate
Molecular Weight (g/mol) 230.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.