Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium 1 decanesulfonate 100G
Catalog No. 502914604
Encompass
Sodium 1-decanesulfonate is an ion-associating reagent.
Catalog No. 50-291-4604 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D3412100G
Sodium 1-decanesulfonate is an ion-associating reagent.