Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium 1-decanesulfonate 25G
Catalog No. 502903127
Encompass
Sodium 1-decanesulfonate is an ion-associating reagent.
Catalog No. 50-290-3127 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D341225G
Sodium 1-decanesulfonate is an ion-associating reagent.