Learn More
Medchemexpress LLC Sodium 1-heptanesulf 10g
SDP
Catalog No. 5000204760
Encompass_Preferred
Sodium 1-heptanesulfonate monohydrate for Ion-Pair Chromatography (Sodium 1-heptanesulfonate monohydrate) is an ion pair reagent for HPLC that can be used to analyze peptides and proteins
Catalog No. 50-002-04760 Supplier Medchemexpress LLC Supplier No. HYW09698910G
Sodium 1-heptanesulfonate monohydrate for Ion-Pair Chromatography (Sodium 1-heptanesulfonate monohydrate) is an ion pair reagent for HPLC that can be used to analyze peptides and proteins