Learn More
Medchemexpress LLC Sodium 1-heptanesulf 25g
SDP
Catalog No. 5000206129
Encompass_Preferred
Sodium 1-heptanesulfonate monohydrate for Ion-Pair Chromatography (Sodium 1-heptanesulfonate monohydrate) is an ion pair reagent for HPLC that can be used to analyze peptides and proteins
Catalog No. 50-002-06129 Supplier Medchemexpress LLC Supplier No. 25GHYW096989
Sodium 1-heptanesulfonate monohydrate for Ion-Pair Chromatography (Sodium 1-heptanesulfonate monohydrate) is an ion pair reagent for HPLC that can be used to analyze peptides and proteins