Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium 1-heptanesulfonate25G
Catalog No. 502899879
Encompass
Sodium 1-heptanesulfonate belongs to the sodium alkylsulfonate group of surfactants. It has a -SO?3 head group with sodium and an alkyl chain attached to the head group. Sodium 1-heptanesulfonate has a critical micelle concentration value of 0.302 mol/dm3.
Catalog No. 50-289-9879 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H276625G
Sodium 1-heptanesulfonate belongs to the sodium alkylsulfonate group of surfactants. It has a -SO?3 head group with sodium and an alkyl chain attached to the head group. Sodium 1-heptanesulfonate has a critical micelle concentration value of 0.302 mol/dm3.