Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium 1-hexanesulfonate monohydrate

Catalog No. 52865250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-865-250G 250 g
52-865-25G 25 g
52-865-50G 50 g
52-865-10G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 52-865-250G Supplier Sigma Aldrich Supplier No. 52865250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium 1-hexanesulfonate monohydrate

Specifications

CAS 207300-91-2
Molecular Formula C6H13NaO3S -+ H2O
Linear Formula CH3(CH2)4 CH2SO3Na -+ H2O
MDL Number MFCD00149549
Quantity 250 g
Synonym 1-Hexanesulfonic acid sodium salt monohydrate
Molecular Weight (g/mol) 206.24
Percent Purity ≥98.0% (T)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.