Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-Naphthaleneacetate 96.0+%, TCI America™
SDP

Catalog No. N000725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N000725G 25 g
N0007500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N000725G Supplier TCI America Supplier No. N000725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61-31-4
Molecular Formula C12H9NaO2
Molecular Weight (g/mol) 208.192
MDL Number MFCD00064967
InChI Key CJUUXVFWKYRHAR-UHFFFAOYSA-M
Synonym 1-Naphthylacetic Acid Sodium Salt, 1-Naphthaleneacetic Acid Sodium Salt, Sodium 1-Naphthylacetate, Na-NAA
PubChem CID 23694671
ChEBI CHEBI:81811
IUPAC Name sodium;2-naphthalen-1-ylacetate
SMILES C1=CC=C2C(=C1)C=CC=C2CC(=O)[O-].[Na+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 208.19
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Naphthaleneacetate
EINECSNumber (4)-0410
RTECSNumber QJ1355000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.