Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium 1-octanesulfonate monohydrate

Catalog No. 7488550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-885-50G 50 g
74-885-10G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74-885-50G Supplier Sigma Aldrich Supplier No. 7488550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium 1-octanesulfonate monohydrate

Specifications

Melting Point ≥300°C (lit.)
CAS 207596-29-0
Quantity 50 g
Molecular Formula C8H17NaO3S -+ H2O
Linear Formula CH3(CH2)6 CH2SO3Na -+ H2O
MDL Number MFCD00149551
Synonym 1-Octanesulfonic acid sodium salt monohydrate
Molecular Weight (g/mol) 234.29
Percent Purity ≥97.0% (T)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.