Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-octanesulfonate monohydrate, ≥99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111013507 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3507 10 g
11-101-3506 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3507 Supplier MilliporeSigma Supelco Supplier No. 7488410G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium 1-octanesulfonate monohydrate (NaOS) is an ion-pairing reagent.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00149551
Synonym 1-Octanesulfonic acid sodium salt monohydrate

Specifications

CAS 207596-29-0
Melting Point ≥300°C
Packaging Poly Bottle
Linear Formula CH3(CH2)6 CH2 SO3 Na · H2O
Quantity 10 g
Molecular Weight (g/mol) 234.29
Formula Weight 234.29
Percent Purity ≥99.0% (T)
Grade For ion pair chromatography
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.