Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium 1-pentanesulfonate monohydrate

Catalog No. 7695510G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-955-10G 10 g
76-955-50G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 76-955-10G Supplier Sigma Aldrich Supplier No. 7695510G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium 1-pentanesulfonate is mainly used as an ion-pairing agent in chromatographic mobile phase.

Specifications

CAS 207605-40-1
Molecular Formula C5H11NaO3S -+ H2O
Linear Formula CH3(CH2)4 SO3Na -+ H2O
MDL Number MFCD00149548
Quantity 10 g
Synonym 1-Pentanesulfonic acid sodium salt
Molecular Weight (g/mol) 192.21
Percent Purity ≥98.0% (T)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.