Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium (+/-)-10-Camphorsulfonate 98.0+%, TCI America™
SDP

Catalog No. C001725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C001725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C001725G Supplier TCI America Supplier No. C001725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34850-66-3
Molecular Formula C10H15NaO4S
Molecular Weight (g/mol) 254.276
MDL Number MFCD00135623
InChI Key AWMAOFAHBPCBHJ-UHFFFAOYSA-M
Synonym sodium camphorsulfonate, sodium 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonate, sodium +/--10-camphorsulfonate, d-camphor-10-sulfonic acid sodium salt, sodium camphorsulfonate anion, bicyclo 2.2.1 heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, sodium salt, +--sodium camphorsulfonate, sodium 2-oxobornane-10-sulphonate, sodium di-10-camphorsulfonate
PubChem CID 23686666
IUPAC Name sodium;(7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonate
SMILES CC1(C2CCC1(C(=O)C2)CS(=O)(=O)[O-])C.[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 254.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium (+/-)-10-Camphorsulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.