Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium [13C]formate | 23102-86-5 | 98.0% | 69.00 g/mol | 13CHNaO2 | 50 MG
SDP

Catalog No. 5000208338
Encompass_Preferred

Formate-13C sodium (Sodium [13C]formate) is an isotopically labeled sodium formate used as a 13C tracer and internal standard for mass spectrometry and metabolic studies. It is supplied as a white to off-white solid with high water solubility and is recommended for cold, moisture-free storage.

  • Isotopically labeled with carbon-13.
  • Purity 98.0%.
  • Molecular weight 69.00 g/mol.
  • Molecular formula 13CHNaO2.
  • Pack size 50 mg.
  • High solubility in water; may require ultrasonic.

Catalog No. 50-002-08338 Supplier Medchemexpress LLC Supplier No. HYW088065S50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formate-13C sodium (Sodium [13C]formate) is an isotopically labeled sodium formate used as a 13C tracer and internal standard for mass spectrometry and metabolic studies. It is supplied as a white to off-white solid with high water solubility and is recommended for cold, moisture-free storage.

  • Isotopically labeled with carbon-13.
  • Purity 98.0%.
  • Molecular weight 69.00 g/mol.
  • Molecular formula 13CHNaO2.
  • Pack size 50 mg.
  • High solubility in water; may require ultrasonic.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.