Learn More
Medchemexpress LLC Sodium [13C]formate | 23102-86-5 | MFCD00064569 | 98.0% | 69.00 | H13CO2Na | 25 MG
SDP
Catalog No. 5000208640
Encompass_Preferred
13C-labeled sodium formate used as an isotope tracer in metabolic and biochemical assays and as a reducing or deprotonating reagent in catalytic and surface chemistry applications. Solid at ambient temperature; white to off-white in color. Store sealed at -20°C and protect from moisture.
  • Isotope-labeled tracer for 13C metabolic and analytical studies.
  • High measured purity suitable for research applications.
  • Solid form convenient for storage and handling.
  • White to off-white appearance indicates expected quality.
  • Keep sealed and refrigerated at -20°C to preserve integrity.
Catalog No. 50-002-08640 Supplier Medchemexpress LLC Supplier No. 25MGHYW088065S
13C-labeled sodium formate used as an isotope tracer in metabolic and biochemical assays and as a reducing or deprotonating reagent in catalytic and surface chemistry applications. Solid at ambient temperature; white to off-white in color. Store sealed at -20°C and protect from moisture.
  • Isotope-labeled tracer for 13C metabolic and analytical studies.
  • High measured purity suitable for research applications.
  • Solid form convenient for storage and handling.
  • White to off-white appearance indicates expected quality.
  • Keep sealed and refrigerated at -20°C to preserve integrity.