Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium 2-(2-Bromophenyl)-1,1-Difluoroethanesulfinate

Catalog No. ALD00462 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ALD00462 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. ALD00462 Supplier Sigma Aldrich Supplier No. ALD00462500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The following Baran Sulfinate is a part of a toolbox of diversification reagents, which are ideal for the late-stage functionalization of nitrogen-containing heterocycles. The regioselectivity can be effectively tuned by modification of the pH and solvent selection.Radical-Based Regioselective C-H Functionalization of Electron-Deficient Heteroarenes: Scope; Tunability; and Predictability

Specifications

CAS 2226371-98-6
Molecular Formula C13H22N4O3
Linear Formula C13H22N4O3
Quantity 5 mg
Synonym tert-Butyl methyl(2-(N-methyl-1H-pyrazole-1-carboxamido)ethyl)carbamate; SHI-20
Molecular Weight (g/mol) 282.34 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.