Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium 2-(3-(2,6-difluoro-4-(1H-pyrazol-4-yl)phenyl)-3-oxoprop-1-en-1-yl)-4-(1-methyl-1H-pyrazol-4-yl | 2519537-70-1 | 98.9% | 50 MG
SDP

Catalog No. 5000230540
Encompass_Preferred

HOIPIN-8 is a research small-molecule inhibitor of the linear ubiquitin chain assembly complex (LUBAC) used to probe ubiquitination and NF-κB signaling in biochemical and cell-based studies. It is supplied as a sodium salt with a molecular weight of 456.38 g/mol.

  • Molecular weight 456.38 g/mol
  • Molecular formula C23H15F2N4NaO3
  • Potent LUBAC inhibitor with IC50 of 11 nM
  • Soluble in DMSO at 50 mg/mL and formulatable for in vivo dosing
  • Store sealed at -20°C; in solution store at -80°C for long-term stability
  • Suitable for biochemical assays and cell-based research

Catalog No. 50-002-30540 Supplier Medchemexpress LLC Supplier No. HY12288250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HOIPIN-8 is a research small-molecule inhibitor of the linear ubiquitin chain assembly complex (LUBAC) used to probe ubiquitination and NF-κB signaling in biochemical and cell-based studies. It is supplied as a sodium salt with a molecular weight of 456.38 g/mol.

  • Molecular weight 456.38 g/mol
  • Molecular formula C23H15F2N4NaO3
  • Potent LUBAC inhibitor with IC50 of 11 nM
  • Soluble in DMSO at 50 mg/mL and formulatable for in vivo dosing
  • Store sealed at -20°C; in solution store at -80°C for long-term stability
  • Suitable for biochemical assays and cell-based research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.