Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium 2,6-Dichloroindophenolate Hydrate

Catalog No. 11981410G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-981-410G 10 g
11-981-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-981-410G Supplier Sigma Aldrich Supplier No. 11981410G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium 2,6-Dichloroindophenolate Hydrate

Specifications

CAS 1266615-56-8
Molecular Formula C12H6Cl2NNaO2 -+ xH2O
Linear Formula C12H6Cl2NNaO2 -+ xH2O
MDL Number MFCD00150014
Quantity 10 g
Synonym 2,6-Dichloro-N-(4-hydroxyphenyl)-1,4-benzoquinoneimine sodium salt; 2,6-Dichloroindophenol sodium salt hydrate; 2,6-Dichlorophenolindophenol sodium salt hydrate
Molecular Weight (g/mol) 290.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.