Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 2-Amino-1-naphthalenesulfonate 98.0+%, TCI America™
SDP

Catalog No. A03525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A03525G Supplier TCI America Supplier No. A03525G

Chemical Identifiers

CAS 25293-52-1
Molecular Formula C10H8NNaO3S
Molecular Weight (g/mol) 245.228
MDL Number MFCD00070484
InChI Key AZKQWFQWHHSLDU-UHFFFAOYSA-M
Synonym 2-Naphthylamine-1-sulfonic Acid Sodium Salt, Tobias Acid Sodium Salt, 2-Amino-1-naphthalenesulfonic Acid Sodium Salt
PubChem CID 23672343
IUPAC Name sodium;2-aminonaphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C=CC(=C2S(=O)(=O)[O-])N.[Na+]

Specifications

Quantity 5 g
Formula Weight 245.23
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Amino-1-naphthalenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.