Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 2-Amino-4-chloro-5-methylbenzenesulfonate 98.0+%, TCI America™
SDP

Catalog No. A089625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A089625G 25 g
A0896500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A089625G Supplier TCI America Supplier No. A089625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6627-59-4
Molecular Formula C7H8ClNNaO3S
Molecular Weight (g/mol) 244.645
MDL Number MFCD00070551
InChI Key OSSZFTJQTIEJDM-UHFFFAOYSA-N
Synonym 5-Chloro-4-methylaniline-2-sulfonic Acid Sodium Salt, 5-Chloro-p-toluidine-2-sulfonic Acid Sodium Salt, Sodium 4-Amino-2-chlorotoluene-5-sulfonate, 4-Amino-2-chlorotoluene-5-sulfonic Acid Sodium Salt, Sodium 5-Chloro-4-methylaniline-2-sulfonate, Sodium 5-
PubChem CID 24187155
IUPAC Name 2-amino-4-chloro-5-methylbenzenesulfonic acid;sodium
SMILES CC1=CC(=C(C=C1Cl)N)S(=O)(=O)O.[Na]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 243.64
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Amino-4-chloro-5-methylbenzenesulfonate
EINECSNumber (3)-2024
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.