Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium 2-bromoethanesulfonate | 4263-52-9 | MFCD00007530 | 97.0% | 211.01 | C2H4BrNaO3S | 100 G
SDP

Catalog No. 5000304155
Encompass_Preferred

Sodium 2-bromoethanesulfonate is a sodium salt used as a laboratory reagent in life-science research and chemical synthesis. It is supplied as a solid and in solution formats and is accompanied by analytical documentation for research use only.

  • Chemical formula C2H4BrNaO3S; CAS 4263-52-9.
  • Molecular weight 211.01 g/mol.
  • Physical state solid; also available in solution formats.
  • Purity typically >97.0%; assay values reported up to 99.7%.
  • Available in multiple package sizes including 100 g and larger quantities by quote.
  • Certificate of analysis and safety data sheet available.
  • Intended for research use only; not for human or diagnostic use.

Catalog No. 50-003-04155 Supplier Medchemexpress LLC Supplier No. HYW014355100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium 2-bromoethanesulfonate is a sodium salt used as a laboratory reagent in life-science research and chemical synthesis. It is supplied as a solid and in solution formats and is accompanied by analytical documentation for research use only.

  • Chemical formula C2H4BrNaO3S; CAS 4263-52-9.
  • Molecular weight 211.01 g/mol.
  • Physical state solid; also available in solution formats.
  • Purity typically >97.0%; assay values reported up to 99.7%.
  • Available in multiple package sizes including 100 g and larger quantities by quote.
  • Certificate of analysis and safety data sheet available.
  • Intended for research use only; not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.