Learn More
Medchemexpress LLC Sodium 2-bromoethanesulfonate | 4263-52-9 | MFCD00007530 | 97.0% | 211.01 | C2H4BrNaO3S | 25 G
SDP
Catalog No. 5000304113
Encompass_Preferred
Sodium 2-bromoethanesulfonate is an organosulfonate reagent supplied as a white solid for laboratory research. It is commonly used in biochemical studies, including inhibition of methanogenesis, and is accompanied by manufacturer documentation and safety data for proper handling and storage.
  • acts as a methanogenesis inhibitor in anaerobic studies.
  • water-soluble sodium sulfonate salt, suitable for aqueous reactions.
  • supplied as a white solid with reported purity and assay information.
  • provided with safety data and handling guidance for laboratory use.
  • packaged in research-scale quantities for convenient use.
Catalog No. 50-003-04113 Supplier Medchemexpress LLC Supplier No. 25GHYW014355
Sodium 2-bromoethanesulfonate is an organosulfonate reagent supplied as a white solid for laboratory research. It is commonly used in biochemical studies, including inhibition of methanogenesis, and is accompanied by manufacturer documentation and safety data for proper handling and storage.
  • acts as a methanogenesis inhibitor in anaerobic studies.
  • water-soluble sodium sulfonate salt, suitable for aqueous reactions.
  • supplied as a white solid with reported purity and assay information.
  • provided with safety data and handling guidance for laboratory use.
  • packaged in research-scale quantities for convenient use.