Learn More
Medchemexpress LLC Sodium 2-bromoethanesulfonate | 4263-52-9 | MFCD00007530 | 97.0% | 211.01 g/mol | C2H4BrNaO3S | 250 G
SDP
Catalog No. 5000302355
Encompass_Preferred
Sodium 2-bromoethanesulfonate is the sodium salt of 2-bromoethanesulfonic acid used as a research reagent in life-science and biochemical applications. It serves as a methanogenesis inhibitor in microbiology studies and as a chemical building block in organic synthesis, and is available in solid and solution forms for laboratory use.
  • Research reagent suitable for biochemical and microbiological studies.
  • Available as solid powder and pre-made solutions for flexibility.
  • Multiple pack sizes accommodate small- and large-scale experiments.
  • High purity and assay data provided for analytical confidence.
  • Molecular formula C2H4BrNaO3S and molecular weight 211.01.
Catalog No. 50-003-02355 Supplier Medchemexpress LLC Supplier No. 250GHYW014355
Sodium 2-bromoethanesulfonate is the sodium salt of 2-bromoethanesulfonic acid used as a research reagent in life-science and biochemical applications. It serves as a methanogenesis inhibitor in microbiology studies and as a chemical building block in organic synthesis, and is available in solid and solution forms for laboratory use.
  • Research reagent suitable for biochemical and microbiological studies.
  • Available as solid powder and pre-made solutions for flexibility.
  • Multiple pack sizes accommodate small- and large-scale experiments.
  • High purity and assay data provided for analytical confidence.
  • Molecular formula C2H4BrNaO3S and molecular weight 211.01.