Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium 2-bromoethanesulfonate | 4263-52-9 | MFCD00007530 | 99.7% | 211.01 g/mol | C2H4BrNaO3S | 500 G
SDP

Catalog No. 5000304065
Encompass_Preferred

Sodium 2-bromoethanesulfonate (CAS 4263-52-9) is an organic sulfonate salt supplied as a white to off-white solid for biochemical and life-science research. The material is accompanied by a certificate of analysis indicating high purity and is intended for research use only; not for human or veterinary applications.

  • High purity (99.7%) per certificate of analysis.
  • White to off-white solid physical form.
  • Molecular formula C2H4BrNaO3S.
  • Provided with COA and SDS documentation.
  • Storage: sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.

Catalog No. 50-003-04065 Supplier Medchemexpress LLC Supplier No. HYW014355500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium 2-bromoethanesulfonate (CAS 4263-52-9) is an organic sulfonate salt supplied as a white to off-white solid for biochemical and life-science research. The material is accompanied by a certificate of analysis indicating high purity and is intended for research use only; not for human or veterinary applications.

  • High purity (99.7%) per certificate of analysis.
  • White to off-white solid physical form.
  • Molecular formula C2H4BrNaO3S.
  • Provided with COA and SDS documentation.
  • Storage: sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.