Learn More
Medchemexpress LLC Sodium 2-ethylhexyl sulfate | 126-92-1 | MFCD00042047 | 40.4% | 232.27 g/mol | C8H17NaO4S | 100 G
SDP
Catalog No. 5000282928
Encompass_Preferred
Sodium 2-ethylhexyl sulfate is an anionic surfactant supplied as a 40% aqueous solution, intended for use as a detergent, wetting agent, and emulsifier in formulation and industrial applications. It reduces surface tension and enhances cleaning performance in a range of products and processes.
  • Anionic surfactant for detergency and emulsification.
  • 40% aqueous formulation suitable for formulation use.
  • Improves wetting and reduces surface tension.
  • Applicable to personal care, household cleaning, and textile processing.
  • Supplied in laboratory-scale pack sizes for testing and formulation development.
Catalog No. 50-002-82928 Supplier Medchemexpress LLC Supplier No. HYW130648100G
Sodium 2-ethylhexyl sulfate is an anionic surfactant supplied as a 40% aqueous solution, intended for use as a detergent, wetting agent, and emulsifier in formulation and industrial applications. It reduces surface tension and enhances cleaning performance in a range of products and processes.
  • Anionic surfactant for detergency and emulsification.
  • 40% aqueous formulation suitable for formulation use.
  • Improves wetting and reduces surface tension.
  • Applicable to personal care, household cleaning, and textile processing.
  • Supplied in laboratory-scale pack sizes for testing and formulation development.