Learn More
Medchemexpress LLC Sodium 2-ethylhexyl sulfate | 126-92-1 | MFCD00042047 | 40.4% | 232.27 g/mol | C8H17NaO4S | 500 G
SDP
Catalog No. 5000283431
Encompass_Preferred
Sodium 2-ethylhexyl sulfate is an anionic surfactant supplied as an aqueous solution for laboratory and formulation use. It serves as a detergent, wetting agent, and emulsifier, and is provided as a colorless to light yellow liquid with an assay around 40%.
  • Anionic surfactant suitable for detergent and emulsifier applications.
  • Supplied as a 40% in water solution (assay ≈ 40.36%).
  • Colorless to light yellow liquid.
  • Commonly used as a wetting agent and emulsifier in formulations and lab procedures.
  • SDS available for handling and safety information.
Catalog No. 50-002-83431 Supplier Medchemexpress LLC Supplier No. HYW130648500G
Sodium 2-ethylhexyl sulfate is an anionic surfactant supplied as an aqueous solution for laboratory and formulation use. It serves as a detergent, wetting agent, and emulsifier, and is provided as a colorless to light yellow liquid with an assay around 40%.
  • Anionic surfactant suitable for detergent and emulsifier applications.
  • Supplied as a 40% in water solution (assay ≈ 40.36%).
  • Colorless to light yellow liquid.
  • Commonly used as a wetting agent and emulsifier in formulations and lab procedures.
  • SDS available for handling and safety information.