Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 2-Naphthol-7-sulfonate Hydrate 98.0+%, TCI America™
SDP

Catalog No. N003625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N003625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N003625G Supplier TCI America Supplier No. N003625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 135-55-7
Molecular Formula C10H7NaO4S
Molecular Weight (g/mol) 246.212
MDL Number MFCD00021625
InChI Key IVXSQYUPCUOMRV-UHFFFAOYSA-M
Synonym Cassella′Cs Acid Sodium Salt, F Acid Sodium Salt, 7-Hydroxy-2-naphthalenesulfonic Acid Sodium Salt, Sodium 7-Hydroxy-2-naphthalenesulfonate, 2-Naphthol-7-sulfonic Acid Sodium Salt
PubChem CID 23667647
IUPAC Name sodium;7-hydroxynaphthalene-2-sulfonate
SMILES C1=CC(=CC2=C1C=CC(=C2)S(=O)(=O)[O-])O.[Na+]

Specifications

Quantity 25 g
Formula Weight 246.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Naphthol-7-sulfonate Hydrate
EINECSNumber (4)-0479
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.