Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium 2-oxo-3-(phenyl-d5)propanoate | 97.1% | 191.17 | C9H2D5NaO3 | 1 MG
SDP

Catalog No. 5000302173
Encompass_Preferred

Phenylpyruvic acid-d5 sodium is the deuterium-labeled sodium salt of phenylpyruvic acid supplied for analytical and research use. It functions as a stable isotopic tracer and internal standard for quantitative analyses, particularly in mass spectrometry and NMR workflows.

  • Deuterium-labeled for use as an isotopic tracer.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • High purity as determined by HPLC.
  • Available in small, accurately weighed pack sizes for analytical workflows.
  • Molecular weight 191.17 and formula C9H2D5NaO3.

Catalog No. 50-003-02173 Supplier Medchemexpress LLC Supplier No. HYW016203S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenylpyruvic acid-d5 sodium is the deuterium-labeled sodium salt of phenylpyruvic acid supplied for analytical and research use. It functions as a stable isotopic tracer and internal standard for quantitative analyses, particularly in mass spectrometry and NMR workflows.

  • Deuterium-labeled for use as an isotopic tracer.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • High purity as determined by HPLC.
  • Available in small, accurately weighed pack sizes for analytical workflows.
  • Molecular weight 191.17 and formula C9H2D5NaO3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.