Learn More
Medchemexpress LLC Sodium 2-oxopropanoate-13C | 124052-04-6 | 98.9% | 111.04 g/mol | C2(13C)CH3NaO3 | 250 MG
SDP
Catalog No. 5000302349
Encompass_Preferred
Sodium 2-oxopropanoate-13C is the 13C-labeled form of sodium pyruvate intended for use as a metabolic tracer and reactive oxygen species (ROS) research reagent. It is supplied as a solid with confirmed isotopic enrichment and HPLC-verified chemical purity; store dry at room temperature, or in solution at -20°C to -80°C for extended storage.
  • 13C isotopic labeling for tracer and quantitation applications.
  • High isotopic enrichment, approximately 99%.
  • High chemical purity, 98.9% by HPLC.
  • Solid, white to off-white appearance for easy handling.
  • Stable storage profile for long-term solutions at low temperatures.
  • For research use only.
Catalog No. 50-003-02349 Supplier Medchemexpress LLC Supplier No. 250MGHYW015913S3
Sodium 2-oxopropanoate-13C is the 13C-labeled form of sodium pyruvate intended for use as a metabolic tracer and reactive oxygen species (ROS) research reagent. It is supplied as a solid with confirmed isotopic enrichment and HPLC-verified chemical purity; store dry at room temperature, or in solution at -20°C to -80°C for extended storage.
  • 13C isotopic labeling for tracer and quantitation applications.
  • High isotopic enrichment, approximately 99%.
  • High chemical purity, 98.9% by HPLC.
  • Solid, white to off-white appearance for easy handling.
  • Stable storage profile for long-term solutions at low temperatures.
  • For research use only.