Learn More
Medchemexpress LLC Sodium 2-oxopropanoate (sodium pyruvate) | 113-24-6 | MFCD00002586 | 99.9% | 110.04 g/mol | C3H3NaO3 | 10 G
SDP
Catalog No. 5000300561
Encompass_Preferred
Sodium 2-oxopropanoate (sodium pyruvate) is the sodium salt of pyruvic acid, a three-carbon metabolite and glycolysis intermediate. Supplied as a high-purity solid, it is used as a biochemical reagent, cell culture supplement, and reactive oxygen species scavenger in metabolic and antioxidant research.
  • High purity (99.86%) suitable for research applications.
  • Solid powder, easy to weigh and dissolve for solution preparation.
  • Compatible with cell culture media as an energy source and supplement.
  • Acts as a reactive oxygen species scavenger for antioxidant studies.
  • Available in multiple pack sizes to support small- and large-scale experiments.
Catalog No. 50-003-00561 Supplier Medchemexpress LLC Supplier No. HYW01591310G
Add to Cart
Add to Cart
Sodium 2-oxopropanoate (sodium pyruvate) is the sodium salt of pyruvic acid, a three-carbon metabolite and glycolysis intermediate. Supplied as a high-purity solid, it is used as a biochemical reagent, cell culture supplement, and reactive oxygen species scavenger in metabolic and antioxidant research.
  • High purity (99.86%) suitable for research applications.
  • Solid powder, easy to weigh and dissolve for solution preparation.
  • Compatible with cell culture media as an energy source and supplement.
  • Acts as a reactive oxygen species scavenger for antioxidant studies.
  • Available in multiple pack sizes to support small- and large-scale experiments.