Learn More
Medchemexpress LLC Sodium 2-oxopropanoate (sodium pyruvate) | 113-24-6 | MFCD00002586 | 99.9% | 110.04 g/mol | C3H3NaO3 | 100 G
SDP
Catalog No. 5000300988
Encompass_Preferred
Sodium 2-oxopropanoate (sodium pyruvate) is the sodium salt of pyruvic acid, provided as a high-purity solid reagent for research and analytical use. It is a three-carbon glycolysis intermediate commonly used as an energy source in cell culture media and as a scavenger of reactive oxygen species in biochemical assays.
  • High reported purity: 99.86%.
  • Molecular formula C3H3NaO3 and molecular weight 110.04 g/mol.
  • Water-soluble solid suitable for solution preparation.
  • Available in multiple pack sizes, including 100 g; larger quantities by quote.
  • Suitable as an analytical standard or reagent in biochemical assays.
Catalog No. 50-003-00988 Supplier Medchemexpress LLC Supplier No. HYW015913100G
Sodium 2-oxopropanoate (sodium pyruvate) is the sodium salt of pyruvic acid, provided as a high-purity solid reagent for research and analytical use. It is a three-carbon glycolysis intermediate commonly used as an energy source in cell culture media and as a scavenger of reactive oxygen species in biochemical assays.
  • High reported purity: 99.86%.
  • Molecular formula C3H3NaO3 and molecular weight 110.04 g/mol.
  • Water-soluble solid suitable for solution preparation.
  • Available in multiple pack sizes, including 100 g; larger quantities by quote.
  • Suitable as an analytical standard or reagent in biochemical assays.