Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 2-Propylvalerate 98.0+%, TCI America™
SDP

Catalog No. S089425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S089425G 25 g
S0894100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S089425G Supplier TCI America Supplier No. S089425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1069-66-5
Molecular Formula C8H15NaO2
Molecular Weight (g/mol) 166.196
MDL Number MFCD00078604
InChI Key AEQFSUDEHCCHBT-UHFFFAOYSA-M
Synonym sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerin
PubChem CID 16760703
ChEBI CHEBI:9925
IUPAC Name sodium;2-propylpentanoate
SMILES CCCC(CCC)C(=O)[O-].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 166.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Propylvalerate
EINECSNumber (2)-0611
RTECSNumber YV7876000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.