Learn More
Medchemexpress LLC Sodium (2S,5R)-2-cyano-7-oxo-1,6-diazabicyclo[3.2.1]octan-6-yl sulfate | 1804915-68-1 | >98.0% | 269.21 | C7H8N3NaO5S | 50MG
SDP
Catalog No. 5000235058
Encompass_Preferred
WCK-4234 sodium (CAS 1804915-68-1) is a diazabicyclooctane (DBO)-class β-lactamase inhibitor supplied as the sodium salt for preclinical research. It exhibits broad-spectrum inhibition of class A, C, and D β-lactamases, including OXA-type carbapenemases, and is used to restore carbapenem efficacy against multidrug-resistant Gram-negative bacteria in pharmacology and microbiology studies.
  • Potent inhibitor of class A, C, and D β-lactamases.
  • Potentiates carbapenem antibiotics in preclinical models.
  • High purity suitable for research applications (>98.0%).
  • Provided as the sodium salt for improved solubility.
  • Well characterized in pharmacokinetic and microbiological studies.
  • Chemical formula C7H8N3NaO5S; molecular weight 269.21.
Catalog No. 50-002-35058 Supplier Medchemexpress LLC Supplier No. HY12560450MG
WCK-4234 sodium (CAS 1804915-68-1) is a diazabicyclooctane (DBO)-class β-lactamase inhibitor supplied as the sodium salt for preclinical research. It exhibits broad-spectrum inhibition of class A, C, and D β-lactamases, including OXA-type carbapenemases, and is used to restore carbapenem efficacy against multidrug-resistant Gram-negative bacteria in pharmacology and microbiology studies.
  • Potent inhibitor of class A, C, and D β-lactamases.
  • Potentiates carbapenem antibiotics in preclinical models.
  • High purity suitable for research applications (>98.0%).
  • Provided as the sodium salt for improved solubility.
  • Well characterized in pharmacokinetic and microbiological studies.
  • Chemical formula C7H8N3NaO5S; molecular weight 269.21.