Learn More
Medchemexpress LLC Sodium 3-methyl-2-oxobutanoate-d7 | 2483831-46-3 | 145.14 | 5MG
SDP
Catalog No. 5000446602
Encompass_Preferred
Sodium 3-methyl-2-oxobutanoate-d7 is the deuterium-labeled form of Sodium 3-methyl-2-oxobutanoate. This compound acts as a precursor of pantothenic acid in Escherichia coli and functions as an endogenous metabolite and metabolic enzyme/protease. The incorporation of stable heavy isotopes, such as deuterium, into drug molecules is primarily used as tracers for quantitation during drug development. Deuteration also shows potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Precursor of pantothenic acid in Escherichia coli
  • Acts as an endogenous metabolite
  • Functions as a metabolic enzyme/protease
  • Used as a tracer in drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-46602 Supplier Medchemexpress LLC Supplier No. HYW006057AS185MG
Sodium 3-methyl-2-oxobutanoate-d7 is the deuterium-labeled form of Sodium 3-methyl-2-oxobutanoate. This compound acts as a precursor of pantothenic acid in Escherichia coli and functions as an endogenous metabolite and metabolic enzyme/protease. The incorporation of stable heavy isotopes, such as deuterium, into drug molecules is primarily used as tracers for quantitation during drug development. Deuteration also shows potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Precursor of pantothenic acid in Escherichia coli
  • Acts as an endogenous metabolite
  • Functions as a metabolic enzyme/protease
  • Used as a tracer in drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs