Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 3-(N-Ethyl-3-methoxyanilino)-2-hydroxy-1-propanesulfonate 98.0+%, TCI America™
SDP

Catalog No. S08261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08261G 1 g
S0826200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08261G Supplier TCI America Supplier No. S08261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82692-96-4
Molecular Formula C12H22NNaO7S
Molecular Weight (g/mol) 347.36
MDL Number MFCD09261431
InChI Key PNGLEYLFMHGIQO-UHFFFAOYNA-M
Synonym 3-(N-Ethyl-3-methoxyanilino)-2-hydroxy-1-propanesulfonic Acid Sodium Salt, N-Ethyl-N-(2-hydroxy-3-sulfopropyl)-3-methoxyaniline Sodium Salt, ADOS
PubChem CID 71307074
IUPAC Name sodium 3-[ethyl(3-methoxyphenyl)amino]-2-hydroxypropane-1-sulfonate dihydrate
SMILES O.O.[Na+].CCN(CC(O)CS([O-])(=O)=O)C1=CC(OC)=CC=C1

Specifications

Melting Point 144°C
Color White
Quantity 1 g
Formula Weight 311.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-(N-Ethyl-3-methoxyanilino)-2-hydroxy-1-propanesulfonate [for Biochemical Research]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.