Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium 3-nitrobenzenesulfonate

Catalog No. 225193100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-519-3100G 100 g
22-519-3500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-519-3100G Supplier Sigma Aldrich Supplier No. 225193100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium 3-nitrobenzenesulfonate (3-Nitrobenzenesulfonic acid sodium salt) was used in the synthesis of quinoline.

Specifications

CAS 127-68-4
Molecular Formula C6H4NNaO5S
Linear Formula O2NC6H4SO3Na
MDL Number MFCD00007490
Quantity 100 g
Synonym 3-Nitrobenzenesulfonic acid sodium salt
Molecular Weight (g/mol) 225.15
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.