Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 4,6-Dihydroxynaphthalene-2-sulfonate 80.0+%, TCI America™
SDP

Catalog No. D2048500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2048500G 500 g
D204825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2048500G Supplier TCI America Supplier No. D2048500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 83732-66-5
Molecular Formula C10H7NaO5S
Molecular Weight (g/mol) 262.211
MDL Number MFCD00142572
InChI Key KJIKTXVDOGQVIJ-UHFFFAOYSA-M
Synonym Sodium 1,7-Dihydroxynaphthalene-3-sulfonate, 4,6-Dihydroxynaphthalene-2-sulfonic Acid Sodium Salt
PubChem CID 5001507
IUPAC Name sodium;4,6-dihydroxynaphthalene-2-sulfonate
SMILES C1=CC(=CC2=C(C=C(C=C21)S(=O)(=O)[O-])O)O.[Na+]

Specifications

Quantity 500 g
Formula Weight 262.21
Percent Purity ≥80.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4,6-Dihydroxynaphthalene-2-sulfonate
EINECSNumber (4)-0512
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.