Learn More
Medchemexpress LLC Sodium 4-aminobenzoate | 555-06-6 | 99.96% | 159.12 | 5 G
SDP
Catalog No. 5000451663
Encompass_Preferred
Sodium 4-aminobenzoate is a biochemical assay reagent. Based on its structure, it displays antioxidant, anticoagulant, fibrinolytic, and immunomodulating activities. It can also be utilized as a protective agent against UV-irradiation and in diagnostic tests for the state of the gastrointestinal tract.
  • Displays antioxidant activity
  • Displays anticoagulant activity
  • Displays fibrinolytic activity
  • Displays immunomodulating activity
  • Protective agent against UV-irradiation
  • Used in diagnostic tests for the gastrointestinal tract
Catalog No. 50-004-51663 Supplier Medchemexpress LLC Supplier No. HYW0101695G
Add to Cart
Add to Cart
Sodium 4-aminobenzoate is a biochemical assay reagent. Based on its structure, it displays antioxidant, anticoagulant, fibrinolytic, and immunomodulating activities. It can also be utilized as a protective agent against UV-irradiation and in diagnostic tests for the state of the gastrointestinal tract.
  • Displays antioxidant activity
  • Displays anticoagulant activity
  • Displays fibrinolytic activity
  • Displays immunomodulating activity
  • Protective agent against UV-irradiation
  • Used in diagnostic tests for the gastrointestinal tract