Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium (4-Chlorophenyl)Methanesulfinate

Catalog No. ALD00468 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ALD00468 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. ALD00468 Supplier Sigma Aldrich Supplier No. ALD00468500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium (4-chlorophenyl)methanesulfinate is an alkyl Baran Diversinate. These reagents facilitate the functionalization of heteroaromatic C-H bonds under simple reaction conditions.The following Baran Sulfinate is a part of a toolbox of diversification reagents, which are ideal for the late-stage functionalization of nitrogen-containing heterocycles. The regioselectivity can be effectively tuned by modification of the pH and solvent selection.Diversinates(TM): Any-Stage Functionalization of (Hetero)aromatic ScaffoldsDiversinates is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 1622426-24-7
Molecular Formula C9H13N3O
Linear Formula C9H13N3O
Quantity 5 mg
Synonym Piperidin-1-yl(1H-pyrazol-1-yl)methanone; SHI-11
Molecular Weight (g/mol) 179.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.