Learn More
Medchemexpress LLC HY-15286 10mg Medchemexpress, Sodium 4-pentynoate CAS:101917-30-0 Purity:>98%
SDP
Catalog No. 502029111
Encompass_Preferred
Medchemexpress, HY-15286 10mg Sodium 4-pentynoate CAS:101917-30-0 Sodium 4-pentynoate is a alkynylacetate analogue, can be metabolically incorporated onto cellular proteins through biosynthetic mechanisms for profiling of acetylated proteins in diverse cell types. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9111 Supplier Medchemexpress LLC Supplier No. HY1528610MG
Medchemexpress, HY-15286 10mg Sodium 4-pentynoate CAS:101917-30-0 Sodium 4-pentynoate is a alkynylacetate analogue, can be metabolically incorporated onto cellular proteins through biosynthetic mechanisms for profiling of acetylated proteins in diverse cell types. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.