Learn More
Medchemexpress LLC HY-15654 100mg Medchemexpress, Sodium 4-phenylbutyrate CAS:1716-12-7 Purity:>98%
SDP
Catalog No. 502081765
Encompass_Preferred
Medchemexpress, HY-15654 100mg Sodium 4-phenylbutyrate CAS:1716-12-7 Sodium 4-phenylbutyrate (4-PBA sodium) is an inhibitor of HDAC and endoplasmic reticulum (ER) stress, used in cancer and infection research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1765 Supplier Medchemexpress LLC Supplier No. HY15654100MG
Medchemexpress, HY-15654 100mg Sodium 4-phenylbutyrate CAS:1716-12-7 Sodium 4-phenylbutyrate (4-PBA sodium) is an inhibitor of HDAC and endoplasmic reticulum (ER) stress, used in cancer and infection research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.