Learn More
Cayman Chemical SodIum 4PhenylbutyrATd11 1mg
SDP
Catalog No. 502987665
Encompass_Preferred
An internal standard for the quantification of sodium 4-phenylbutyrate by GC- or LC-MS
Catalog No. 50-298-7665 Supplier Cayman Chemical Supplier No. 220851 MG
An internal standard for the quantification of sodium 4-phenylbutyrate by GC- or LC-MS