Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium 5-[(3-ethynylphenyl)amino]pyrimido[4,5-c]quinoline-8-carboxylate | 1333382-30-1 | MFCD26940344 | 99.5% | 362.32 | C20H11N4NaO2 | 10 MG
SDP

Catalog No. 5000298667
Encompass_Preferred

CX-5011 is a small-molecule research compound that functions as a casein kinase 2 (CK2) inhibitor. Presented as the sodium salt, it has been reported to activate Rac1 and induce apoptosis in cancer cell lines and is intended for research use only.

  • Casein kinase 2 (CK2) inhibitor activity
  • Induces Rac1 activation and apoptosis in cancer cells
  • Molecular weight 362.32; chemical formula C20H11N4NaO2
  • High purity by HPLC (typical of research-grade reagents)
  • Storage: powder at -20°C for long-term; in solvent keep at -80°C when possible

Catalog No. 50-002-98667 Supplier Medchemexpress LLC Supplier No. HY50733A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CX-5011 is a small-molecule research compound that functions as a casein kinase 2 (CK2) inhibitor. Presented as the sodium salt, it has been reported to activate Rac1 and induce apoptosis in cancer cell lines and is intended for research use only.

  • Casein kinase 2 (CK2) inhibitor activity
  • Induces Rac1 activation and apoptosis in cancer cells
  • Molecular weight 362.32; chemical formula C20H11N4NaO2
  • High purity by HPLC (typical of research-grade reagents)
  • Storage: powder at -20°C for long-term; in solvent keep at -80°C when possible

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.