Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium AA Standard, 1000 ppm Na in 3% HCl, Ricca Chemical
SDP

Catalog No. ANA1KH100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ANA1KH-100 100 mL Natural LDPE Bottle
ANA1KH-500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ANA1KH-100 Supplier Ricca Chemical Company Supplier No. ANA1KH100
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5 , 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Na
CAS Min % 95.0, 3.05, 0.22
CAS Max % 99.0, 3.18, 0.23
Equivalence 1 mL = 1 mg Na
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 100 mL
Chemical Name or Material Sodium AA Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.