Learn More
Chata Biosystems Sodium Acetate, 0.05M, pH 4.5 - 100L CHEM+NECT
Catalog No. NC9979187
Encompass
Our CHEM+NECT™ system is designed to provide a closed system delivery option of introducting reagents into a clean and microbiological-controlled process. Designed to provide a closed system delivery option of introducing reagents into a clean and microbiological-controlled process. The CHEM+NECTTM bag is a sanitary and more environmentally sustainable alternative to glass and plastic bottles. The CHEM+NECTTM bag is ideal for processes in which pumping minor volumes of reagents and chemicals is used.
Catalog No. NC9979187 Supplier Chata Biosystems Supplier No. 56444
Our CHEM+NECT™ system is designed to provide a closed system delivery option of introducting reagents into a clean and microbiological-controlled process. Designed to provide a closed system delivery option of introducing reagents into a clean and microbiological-controlled process. The CHEM+NECTTM bag is a sanitary and more environmentally sustainable alternative to glass and plastic bottles. The CHEM+NECTTM bag is ideal for processes in which pumping minor volumes of reagents and chemicals is used.