Learn More
Medchemexpress LLC Sodium acetate-1,2-13C2 | 56374-56-2 | MFCD00037667 | 98.6% | 84.02 | 13C2H3NaO2 | 25mg
SDP
Catalog No. 5000270295
Encompass_Preferred
Sodium Acetate-1 2-13C2 is a stable isotope labeled compound with biomarker and metabolic tracking activity Sodium Acetate-1 2-13C2 is often used in biochemical research for the analysis of metabolic pathways Sodium Acetate-1 2-13C2 is also used in medical imaging and compound development to improve the understanding of biological processes
Catalog No. 50-002-70295 Supplier Medchemexpress LLC Supplier No. HYW74920925MG
Sodium Acetate-1 2-13C2 is a stable isotope labeled compound with biomarker and metabolic tracking activity Sodium Acetate-1 2-13C2 is often used in biochemical research for the analysis of metabolic pathways Sodium Acetate-1 2-13C2 is also used in medical imaging and compound development to improve the understanding of biological processes