Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate, 2.00 M (2.00 N), Ricca Chemical
SDP

APHA for Zinc Testing, EPA for Fluoride Testing

Supplier:  Ricca Chemical Company 713032

 View more versions of this product

Catalog No. 7130-32


Add to Cart

Description

Description

Specifications

Specifications

Sodium Acetate
136.08 g/mol
Sodium Acetate
APHA for Zinc, EPA for Fluoride
1 L
7732-18-5
Laboratory
Natural Poly Bottle
7.5 to 9.2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.